1,3,5-Tri(1H-imidazol-1-yl)benzene 97%

Supplier: Ambeed
Warning

A524563-100MG A524563-10G A524563-250MG A524563-5G A524563-1G
77241-718EA 6 USD
77241-718 77241-720 77241-722 77241-724 77572-670
1,3,5-Tri(1H-imidazol-1-yl)benzene 97%
1,3,5-Tri(1H-imidazol-1-yl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₆
MDL Number: MFCD23103303
CAS Number: 528543-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR