Methyl oxindole-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl Oxindole-5-carboxylate

A524331-1G A524331-250MG A524331-25G A524331-5G A524331-10G
77279-806EA 26.78 USD
77279-806 77279-808 77279-810 77279-812 77749-092
Methyl oxindole-5-carboxylate 95%
Methyl oxindole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.1834 g/mol
MDL Number: MFCD02179600
CAS Number: 199328-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR