1-Butynylbenzene 99%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-1-butyne

A523695-100G A523695-25G A523695-5G
77241-712EA 515.99 USD
77241-712 77241-714 77241-716
1-Butynylbenzene 99%
1-Butynylbenzene

Precaución: for Research Use Only

Formula: C₆H₅CCCH₂CH₃
MDL Number: MFCD00039945
CAS Number: 622-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR