Src inhibitor-1 99%
Supplier: Ambeed
|
Warning
|
A523201-100MG
A523201-10MG
A523201-1G
A523201-1MG
A523201-250MG
A523201-25MG
A523201-50MG
A523201-5MG
77241-690EA
61
USD
77241-690
77241-692
77241-694
77241-696
77241-698
77241-700
77241-702
77241-704
Src inhibitor-1 99%
Src inhibitor-1
Precaución: for Research Use Only
Formula:
C₂₂H₁₉N₃O₃ |
MDL Number:
MFCD01815300 CAS Number: 179248-59-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...