3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol 95%

Supplier: Ambeed
Warning

Synonyms: 3-((tert-Butyldimethylsilyl)oxy)propan-1-ol, 3-[[tert-Butyl(dimethyl)silyl]oxy]propan-1-ol, 3-[(tert-Butyldimethylsilyl)oxy]-1-propanol

A522743-100G A522743-10G A522743-1G A522743-5G A522743-25G
77279-796EA 241.02 USD
77279-796 77279-798 77279-800 77279-802 77572-662
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol 95%
3-[[tert-Butyl(dimethyl)silyl]oxy]-1-propanol

Precaución: for Research Use Only

Formula: C₉H₂₂O₂Si
MW: 190.36 g/mol
MDL Number: MFCD00187968
CAS Number: 73842-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR