[4,4'-Di-t-butyl-2,2'-bipyridine][bis[5-(t-butyl)-2-[4-(t-butyl)-2-pyridinyl-kN]phenyl-kC]iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A522406-100MG A522406-1G A522406-250MG A522406-50MG
77253-510EA 41 USD
77253-510 77253-512 77253-514 77253-516
[4,4'-Di-t-butyl-2,2'-bipyridine][bis[5-(t-butyl)-2-[4-(t-butyl)-2-pyridinyl-kN]phenyl-kC]iridium(III) hexafluorophosphate 97%
[4,4'-Di-t-butyl-2,2'-bipyridine][bis[5-(t-butyl)-2-[4-(t-butyl)-2-pyridinyl-kN]phenyl-kC]iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₅₆H₇₂F₆IrN₄P
MW: 1138.37 g/mol
MDL Number: MFCD28369262
CAS Number: 808142-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR