3,7-Dibromo-10H-phenothiazine 95%

Supplier: Ambeed
Warning

A522338-250MG A522338-1G
77265-930EA 23 USD
77265-930 77617-228
3,7-Dibromo-10H-phenothiazine 95%
3,7-Dibromo-10H-phenothiazine

Precaución: for Research Use Only

Formula: C₁₂H₇Br₂NS
MDL Number: MFCD30531816
CAS Number: 21667-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR