(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-5,5',6,6'-Tetramethyl-3,3'-di-t-butyl-1,1'-biphenyl-2,2'-diol, (S)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, (S)-BIPHEN H2

A522078-100MG A522078-1G A522078-5G
77253-504EA 55 USD
77253-504 77253-506 77253-508
(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol 98%
(S)-(-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol

Precaución: for Research Use Only

Formula: C₂₄H₃₄O₂
MW: 354.526 g/mol
MDL Number: MFCD01862439
CAS Number: 205927-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR