1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene 98%

Supplier: Ambeed
Warning

Synonyms: (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene, 1,2-Bis((2R,5R)-2,5-dimethylphospholan-1-yl)benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholan-1-yl]benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholano]benzene, (R,R)-Me-DUPHOS

A521843-100MG A521843-1G A521843-250MG A521843-5G
77253-498EA 87.55 USD
77253-498 77253-500 77253-502 77747-066
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene 98%
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene

Precaución: for Research Use Only

Formula: C₁₈H₂₈P₂
MW: 306.37 g/mol
MDL Number: MFCD00142336
CAS Number: 147253-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR