tert-Butyl 6-bromo-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A521786-100MG
A521786-250MG
77265-926EA
114.33
USD
77265-926
77265-928
tert-Butyl 6-bromo-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate 97%
tert-Butyl 6-bromo-2-oxospiro[indoline-3,3'-pyrrolidine]-1'-carboxylate
Precaución: for Research Use Only
Formula:
C₁₆H₁₉BrN₂O₃ MW: 367.24 g/mol |
MDL Number:
MFCD29761999 CAS Number: 2089310-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...