2,3-Dibromo-6-chloropyridine 97%

Supplier: Ambeed
Danger

A521742-1G A521742-250MG A521742-25G A521742-5G
77253-490EA 20 USD
77253-490 77253-492 77253-494 77253-496
2,3-Dibromo-6-chloropyridine 97%
2,3-Dibromo-6-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClN
MDL Number: MFCD06798229
CAS Number: 885952-16-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR