3,4-Dihydro-2H-pyran-5-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydro-2H-pyran-5-carboxaldehyde

A521291-100MG A521291-1G A521291-250MG A521291-5G
77279-780EA 15.45 USD
77279-780 77279-782 77279-784 77279-786
3,4-Dihydro-2H-pyran-5-carbaldehyde 98%
3,4-Dihydro-2H-pyran-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00085019
CAS Number: 25090-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR