2'-Methylacetanilide 98%

Supplier: Ambeed
Warning

Synonyms: o-Acetotoluidide, o-Acetotoluidid, N-(o-Tolyl)acetamide, N-(2-Methylphenyl)acetamide

A521073-100G A521073-25G A521073-5G
77279-774EA 59.74 USD
77279-774 77279-776 77279-778
2'-Methylacetanilide 98%
2'-Methylacetanilide

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD00014961
CAS Number: 120-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR