2-Aminopyridine-N-oxide 98%

Supplier: Ambeed
Warning

A520412-250MG A520412-1G
77265-924EA 59 USD
77265-924 77747-065
2-Aminopyridine-N-oxide 98%
2-Aminopyridine N-oxide

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MDL Number: MFCD00059168
CAS Number: 14150-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR