4,4'-Difluorodiphenylacetylene 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(4-fluorophenyl)ethyne

A519954-100MG A519954-1G A519954-250MG A519954-5G
77292-272EA 30.9 USD
77292-272 77292-274 77292-276 77292-278
4,4'-Difluorodiphenylacetylene 97%
4,4'-Difluorodiphenylacetylene

Precaución: for Research Use Only

Formula: C₁₄H₈F₂
MW: 214.22 g/mol
MDL Number: MFCD01596894
CAS Number: 5216-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR