Br-Mmc (=4-bromomethyl-7-methoxycoumarin) 98%

Supplier: Ambeed
Danger

Synonyms: 4-Bromomethyl-7-methoxycoumarin, 4-(Bromomethyl)-7-methoxycoumarin, 4-(Bromomethyl)-7-methoxy-2H-chromen-2-one, Br-Mmc

A519403-1G A519403-250MG A519403-5G A519403-10G
77292-266EA 60.77 USD
77292-266 77292-268 77292-270 77749-082
Br-Mmc (=4-bromomethyl-7-methoxycoumarin) 98%
Br-Mmc (=4-bromomethyl-7-methoxycoumarin)

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₃
MW: 269.09 g/mol
MDL Number: MFCD00006869
CAS Number: 35231-44-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR