2-Amino-4-methylthiazole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A519243-10G A519243-1G A519243-5G
77253-478EA 134.93 USD
77253-478 77253-480 77253-482
2-Amino-4-methylthiazole-5-carboxylic acid 98%
2-Amino-4-methylthiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MW: 158.18 g/mol
MDL Number: MFCD01853938
CAS Number: 67899-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR