3-Fluoro-4-(methoxy(methyl)carbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: 3-Fluoro-4-[methoxy(methyl)carbamoyl]phenylboronic acid, (3-Fluoro-4-(methoxy(methyl)carbamoyl)phenyl)boronic acid

A518965-1G A518965-250MG A518965-5G
77279-766EA 66 USD
77279-766 77279-768 77279-770
3-Fluoro-4-(methoxy(methyl)carbamoyl)phenylboronic acid 98%
3-Fluoro-4-(methoxy(methyl)carbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BFNO₄
MW: 227.0000 g/mol
MDL Number: MFCD08689477
CAS Number: 913835-59-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR