3-Fluoro-4-methoxybenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A518888-100G
A518888-10G
A518888-1G
A518888-25G
A518888-500G
A518888-5G
77241-666EA
51
USD
77241-666
77241-668
77241-670
77241-672
77241-674
77241-676
3-Fluoro-4-methoxybenzoic acid 98%
3-Fluoro-4-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇FO₃ |
MDL Number:
MFCD00060675 CAS Number: 403-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...