2-Methyl-1-pyrroline 98%

Supplier: Ambeed
Danger

A518878-10G A518878-1G A518878-25G A518878-5G
77265-912EA 53.56 USD
77265-912 77265-914 77265-916 77265-918
2-Methyl-1-pyrroline 98%
2-Methyl-1-pyrroline

Precaución: for Research Use Only

Formula: C₅H₉N
MW: 83.13 g/mol
MDL Number: MFCD00128806
CAS Number: 872-32-2
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR