(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl 98%

Supplier: Ambeed
Danger

A518599-1G A518599-250MG
77292-260EA 123 USD
77292-260 77551-558
(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl 98%
(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl

Precaución: for Research Use Only

Formula: C₂₀H₁₅NO
MDL Number: MFCD01882346
CAS Number: 137848-29-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR