2-Bromothioanisole 98%

Supplier: Ambeed
Warning

A518478-100G A518478-1G A518478-25G A518478-500G A518478-5G
77265-902EA 181 USD
77265-902 77265-904 77265-906 77265-908 77265-910
2-Bromothioanisole 98%
2-Bromothioanisole

Precaución: for Research Use Only

Formula: C₇H₇BrS
MDL Number: MFCD00000066
CAS Number: 19614-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR