2,4-Bis(ɑ,ɑ-dimethylbenzyl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Bis(2-phenylpropan-2-yl)phenol, 2,4-Di-ɑ-cumylphenol

A518262-100G A518262-25G A518262-500G
77253-468EA 16.48 USD
77253-468 77253-470 77253-472
2,4-Bis(ɑ,ɑ-dimethylbenzyl)phenol 97%
2,4-Bis(alpha,alpha-dimethylbenzyl)phenol

Precaución: for Research Use Only

Formula: C₂₄H₂₆O
MW: 330.47 g/mol
MDL Number: MFCD00191242
CAS Number: 2772-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR