7-Bromo-1-methylnaphthalene 97%

Supplier: Ambeed
Warning

A518177-250MG A518177-1G
77253-466EA 75 USD
77253-466 77747-064
7-Bromo-1-methylnaphthalene 97%
7-Bromo-1-methylnaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉Br
MDL Number: MFCD17012268
CAS Number: 33295-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR