NPS-1034 98+%

Supplier: Ambeed
Warning

Synonyms: N-(3-Fluoro-4-((3-phenyl-1H-pyrrolo[2,3-b]pyridin-4-yl)oxy)phenyl)-2-(4-fluorophenyl)-1,5-dimethyl-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxamide

A518169-10MG A518169-1MG A518169-50MG A518169-5MG
77265-892EA 53.56 USD
77265-892 77265-894 77265-896 77265-898
NPS-1034 98+%
NPS-1034

Precaución: for Research Use Only

Formula: C₃₁H₂₃F₂N₅O₃
MW: 551.54 g/mol
MDL Number: MFCD29905447
CAS Number: 1221713-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR