4-(3,4-Dimethylphenyl)-2,2-dimethyl-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

A517958-100MG A517958-1G A517958-250MG
77241-660EA 39 USD
77241-660 77241-662 77241-664
4-(3,4-Dimethylphenyl)-2,2-dimethyl-4-oxobutanoic acid 97%
4-(3,4-Dimethylphenyl)-2,2-dimethyl-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₃
MDL Number: MFCD09801870
CAS Number: 951894-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR