3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-2-phenylquinolin-4(1H)-one

A516650-100MG A516650-1G A516650-250MG
77279-760EA 60.77 USD
77279-760 77279-762 77279-764
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%
3-Hydroxy-2-phenyl-4(1H)-quinolinone

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO₂
MW: 237.26 g/mol
MDL Number: MFCD00451324
CAS Number: 31588-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR