4-(Trifluoroacetyl)morpholine 97%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoro-1-morpholinoethanone, 4-(2,2,2-Trifluoroethanoyl)morpholine, 1-(Morpholin-4-yl)-2,2,2-trifluoroethan-1-one

A515958-1G A515958-250MG A515958-25G A515958-5G A515958-100G
77292-242EA 6.18 USD
77292-242 77292-244 77292-246 77292-248 77749-067
4-(Trifluoroacetyl)morpholine 97%
4-(Trifluoroacetyl)morpholine

Precaución: for Research Use Only

Formula: C₆H₈F₃NO₂
MW: 183.128 g/mol
MDL Number: MFCD00465461
CAS Number: 360-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR