DMT-2'-O-MOE-rA(Bz) phosphoramidite 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-5-(6-Benzamido-9H-purin-9-yl)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-(2-methoxyethoxy)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite

A515905-5G A515905-1G A515905-250MG
77265-880EA 432.6 USD
77265-880 77572-644 77749-066
DMT-2'-O-MOE-rA(Bz) phosphoramidite 97%
DMT-2'-O-MOE-rA(Bz) phosphoramidite

Precaución: for Research Use Only

Formula: C₅₀H₅₈N₇O₉P
MW: 932.01 g/mol
MDL Number: MFCD29472587
CAS Number: 251647-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR