ABT-639 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Chloro-2-fluoro-N-(2-fluorophenyl)-5-(octahydropyrrolo[1,2-a]pyrazine-2-carbonyl)benzenesulphonamide

A515853-10MG A515853-1MG
77241-650EA 77 USD
77241-650 77241-652 77241-654
ABT-639 98+%
ABT-639

Precaución: for Research Use Only

Formula: C₂₀H₂₀ClF₂N₃O₃S
MW: 455.91 g/mol
MDL Number: MFCD29924737
CAS Number: 1235560-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR