(±)-1-Pentyn-3-ol 95%

Supplier: Ambeed
Danger

Synonyms: (±)-Ethyl ethynyl carbinol, 1-Pentyn-3-ol

A515600-1G A515600-25G A515600-5G
77241-644EA 17.01 USD
77241-644 77241-646 77241-648
(±)-1-Pentyn-3-ol 95%
(±)-1-Pentyn-3-ol

Precaución: for Research Use Only

Formula: C₅H₈O
MDL Number: MFCD00004572
CAS Number: 4187-86-4
UN: 1986
ADR: 3,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR