4-(Fluorosulfonyl)benzoic acid 95%

Supplier: Ambeed
Danger

A515212-100MG A515212-250MG A515212-1G
77292-238EA 29.87 USD
77292-238 77292-240 77594-608
4-(Fluorosulfonyl)benzoic acid 95%
4-(Fluorosulfonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₇H₅FO₄S
MW: 204.18 g/mol
MDL Number: MFCD00007420
CAS Number: 455-26-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR