2,6-Dimethoxytoluene 98%

Supplier: Ambeed
Warning

A514625-25G A514625-5G
77253-438EA 48.41 USD
77253-438 77253-440
2,6-Dimethoxytoluene 98%
2,6-Dimethoxytoluene

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
MDL Number: MFCD00008374
CAS Number: 5673-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR