(S)-5-(Benzyloxy)-2-(2,2-diphenylacetyl)-6-methoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Olodanrigan

A513856-10MG A513856-1MG A513856-25MG A513856-50MG A513856-5MG
77265-848EA 81.37 USD
77265-848 77265-850 77265-852 77265-854 77265-856
(S)-5-(Benzyloxy)-2-(2,2-diphenylacetyl)-6-methoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 97%
(S)-5-(Benzyloxy)-2-(2,2-diphenylacetyl)-6-methoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₃₂H₂₉NO₅
MW: 507.57 g/mol
MDL Number: MFCD28502251
CAS Number: 1316755-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR