4-Chloro-3-fluoroaniline 98%
Supplier: Ambeed
|
Danger
|
A513816-1G
A513816-25G
A513816-5G
77292-222EA
5.15
USD
77292-222
77292-224
77292-226
4-Chloro-3-fluoroaniline 98%
4-Chloro-3-fluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₅ClFN MW: 145.56 g/mol |
MDL Number:
MFCD01090987 CAS Number: 367-22-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...