4,6-Dichloro-1H-pyrazolo[3,4-b]pyridine 98%
Supplier: Ambeed
|
Warning
|
A513782-100MG
A513782-1G
A513782-250MG
A513782-5G
77265-840EA
49.44
USD
77265-840
77265-842
77265-844
77265-846
4,6-Dichloro-1H-pyrazolo[3,4-b]pyridine 98%
4,6-Dichloro-1H-pyrazolo[3,4-b]pyridine
Precaución: for Research Use Only
Formula:
C₆H₃Cl₂N₃ MW: 188.01 g/mol |
MDL Number:
MFCD31580212 CAS Number: 2105905-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...