2,2-Dimethylglutaric anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 3,3-Dimethyldihydro-2H-pyran-2,6(3H)-dione

A513741-1G A513741-250MG A513741-25G A513741-5G
77253-430EA 32.96 USD
77253-430 77253-432 77253-434 77253-436
2,2-Dimethylglutaric anhydride 98%
2,2-Dimethylglutaric anhydride

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.1525 g/mol
MDL Number: MFCD00006682
CAS Number: 2938-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR