4-Isobutylbenzaldehyde 95%

Supplier: Ambeed
Warning

A513178-100G A513178-10G A513178-25G A513178-5G
77292-208EA 152.44 USD
77292-208 77292-210 77292-212 77292-214
4-Isobutylbenzaldehyde 95%
4-Isobutylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MDL Number: MFCD00075760
CAS Number: 40150-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR