4-Bromotetrafluorophenol 96%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,3,5,6-tetrafluorophenol

A512754-1G A512754-250MG A512754-10G A512754-5G
77292-204EA 32.96 USD
77292-204 77292-206 77617-196 77749-057
4-Bromotetrafluorophenol 96%
4-Bromotetrafluorophenol

Precaución: for Research Use Only

Formula: C₆HBrF₄O
MW: 244.98 g/mol
MDL Number: MFCD06246024
CAS Number: 1998-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR