7-Bromo-1,2,3,4-tetrahydronaphthalen-1-ol 97%

Supplier: Ambeed
Warning

A512719-100MG A512719-1G A512719-250MG
77265-834EA 44.29 USD
77265-834 77265-836 77265-838
7-Bromo-1,2,3,4-tetrahydronaphthalen-1-ol 97%
7-Bromo-1,2,3,4-tetrahydronaphthalen-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
MW: 227.09 g/mol
MDL Number: MFCD11850074
CAS Number: 75693-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR