2-Chloroquinoline-6-carbonitrile 95%

Supplier: Ambeed
Warning

A511889-100MG A511889-250MG
77265-822EA 29 USD
77265-822 77265-824
2-Chloroquinoline-6-carbonitrile 95%
2-Chloroquinoline-6-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₅ClN₂
MDL Number: MFCD09038087
CAS Number: 78060-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR