Anthracene-9,10-diyl diacetate 97%

Supplier: Ambeed
Warning

A511879-1G A511879-250MG
77253-424EA 94.76 USD
77253-424 77253-426
Anthracene-9,10-diyl diacetate 97%
Anthracene-9,10-diyl diacetate

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₄
MW: 294.30 g/mol
MDL Number: MFCD00035994
CAS Number: 604-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR