2,3-Dichloro-6-fluorobenzoic acid 98+%
Supplier: Ambeed
|
Warning
|
A510888-1G
A510888-250MG
77253-420EA
86.52
USD
77253-420
77253-422
2,3-Dichloro-6-fluorobenzoic acid 98+%
2,3-Dichloro-6-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃Cl₂FO₂ |
MDL Number:
MFCD00090506 CAS Number: 32890-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...