2-(Trifluoromethoxy)terephthalic acid 95%

Supplier: Ambeed
Warning

A510520-1G A510520-250MG A510520-5G
77253-414EA 117 USD
77253-414 77253-416 77253-418
2-(Trifluoromethoxy)terephthalic acid 95%
2-(Trifluoromethoxy)terephthalic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃O₅
MDL Number: MFCD00203986
CAS Number: 175278-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR