7H-Pyrrolo[2,3-d]pyrimidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A510181-100MG A510181-1G A510181-250MG
77241-612EA 55 USD
77241-612 77241-614 77241-616
7H-Pyrrolo[2,3-d]pyrimidine-2-carboxylic acid 98%
7H-Pyrrolo[2,3-d]pyrimidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD17016001
CAS Number: 1314723-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR