4-Allyl-1-Boc-4-hydroxypiperidine 95%

Supplier: Ambeed
Warning

A510131-1G A510131-250MG A510131-5G
77292-182EA 36.05 USD
77292-182 77292-184 77749-044
4-Allyl-1-Boc-4-hydroxypiperidine 95%
4-Allyl-1-Boc-4-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₃
MW: 241.33 g/mol
MDL Number: MFCD09910310
CAS Number: 203662-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR