7-Cyano-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A509714-100MG A509714-1G A509714-250MG
77265-790EA 77.25 USD
77265-790 77265-792 77265-794
7-Cyano-1H-indole-2-carboxylic acid 98%
7-Cyano-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₂
MW: 186.16 g/mol
MDL Number: MFCD20645477
CAS Number: 796870-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR