Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) complex 98%

Supplier: Ambeed
Warning

A509223-100MG A509223-1G A509223-250MG
77265-784EA 50 USD
77265-784 77265-786 77265-788
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) complex 98%
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) complex

Precaución: for Research Use Only

Formula: C₇₂H₄₈F₆N₆PRu
MW: 1243.23 g/mol
MDL Number: MFCD03791108
CAS Number: 123148-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR