Ethyl 4-bromothieno[2,3-c]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

A509091-1G A509091-250MG
77241-600EA 100.94 USD
77241-600 77241-602
Ethyl 4-bromothieno[2,3-c]pyridine-2-carboxylate 97%
Ethyl 4-bromothieno[2,3-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂S
MW: 286.15 g/mol
MDL Number: MFCD30188747
CAS Number: 1809004-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR