5(6)-Carboxyfluorescein 98% + (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: 5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid

A508933-1G A508933-1MG A508933-250MG A508933-25G A508933-5G
77265-774EA 31.93 USD
77265-774 77265-776 77265-778 77265-780 77265-782
5(6)-Carboxyfluorescein 98% + (mixture of isomers)
5(6)-Carboxyfluorescein

Precaución: for Research Use Only

Formula: C₄₂H₂₄O₁₄
MW: 752.63 g/mol
MDL Number: MFCD00151081
CAS Number: 72088-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR